Order Entry
Belgium
ContactUsLinkComponent
 
Application specific filter papers, grade 287 ½, Whatman™
 
undefined
Application specific filter papers, grade 287 ½, Whatman™

 

  • Catalog No:
  • 512-0581
  • 512-0582
  • Grade:
  • 287 ½
  • 287 ½
  • Weight:
  • 161 g/m²
  • 161 g/m²
  • Thickness:
  • 360 µm
  • 360 µm
  • Filtration speed:
  • 330 s (Herzberg)
  • 330 s (Herzberg)

 

Frequently Bought Together