Order Entry
Belgium
Orders LinkContactUsLinkComponent
Application specific filter papers, grade 72, Whatman™

Application specific filter papers, grade 72, Whatman™

Application specific filter papers, grade 72, Whatman™
Certificates
Application specific filter papers, grade 72, Whatman™
Application specific filter papers, grade 72, Whatman™

3 Options Available Below

Product Details & Documents

Grade 72 activated carbon air filter paper is designed for air pollution monitoring of radioactive iodine in medical and nuclear installations.

  • High-quality composite cellulose/glass filter paper
  • Loaded with activated carbon for radioactive iodine absorption
  • Suitable for use in radioactive environments

Air pollution monitoring of radioactive iodine in medical or nuclear installations is a critical safety requirement. Grade 72 cellulose filter papers are designed specifically for these environments, delivering sensitive and accurate results by absorbing any radioactive iodine in the air for analysis.

Specifications for Product Family

Specifications

  • Catalog No:
  • 512-0579
  • WHAT1872-050
  • WHAT1872-055
  • Grade:
  • 72
  • 72
  • 72
  • Weight:
  • 195 g/m²
  • 195 g/m²
  • 195 g/m²
  • Thickness:
  • 800 µm
  • 800 µm
  • 800 µm

Specifications

Product Family Options

Product Information

  • Grade
    Size
    Availability
    Price
  • 72
    47 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 196,00 €
    196,00 €
    1 * 100 ST
       196,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    512-0579
    Grade
    72
    Weight
    195 g/m²
    Thickness
    800 µm
  • 72
    50 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 212,00 €
    212,00 €
    1 * 100 ST
       212,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    WHAT1872-050
    Grade
    72
    Weight
    195 g/m²
    Thickness
    800 µm
  • 72
    55 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 101,00 €
    101,00 €
    1 * 100 ST
       101,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    WHAT1872-055
    Grade
    72
    Weight
    195 g/m²
    Thickness
    800 µm