Order Entry
Belgium
Orders LinkContactUsLinkComponent
Density hydrometers

Density hydrometers

Supplier:   Ludwig Schneider
Density hydrometers
Supplier:  Ludwig Schneider
Certificates
Density hydrometers
Density hydrometers

2 Options Available Below

Product Details & Documents

Density hydrometers without thermometer calibrated at 20 °C.

Ordering information: A works calibration certificate is available on request.

Packaging: In a square plastic sleeve.

Product Family Options

Product Information

  • Range
    Division
    Length
    Availability
    Price
  • 2,800 - 3,000 g/ml
    0,002 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 72,70 €
    72,70 €
    1 * 1 ST
       72,70 €

  • 3,200 - 3,400 g/ml
    0,002 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 75,30 €
    75,30 €
    1 * 1 ST
       75,30 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.