ContactUsLinkComponent
GC installation kit
GC installation kit
Catalog # REST21325
Supplier:  Restek
undefined
GC installation kit
Catalog # REST21325
Supplier:  Restek
Specifications
  • Description:
    GC installation kit
  • Pk:
    1
Specifications
About this item
This kit contains the tubing and fittings needed to add an additional GC to lab bench.
Delivery: This kit includes tubing cutter, one 3,17×6,35 mm reamer, one 6,35×3,17 mm brass tube end reducer, one 11,11 mm wrench, one 12,7 mm wrench, four 3,17 mm brass tees, ten 3,17 mm brass nuts, ten brass front and back ferrules and 15,2 m of instrument-grade cleaned 3,17 mm copper tubing.