Order Entry
Belgium
ContactUsLinkComponent
Mouse TNFRSF7 ELISA kit
Mouse TNFRSF7 ELISA kit
Catalog # BIRBORB138064-96
Supplier:  Biorbyt
CAS Number:  
undefined
Mouse TNFRSF7 ELISA kit
Catalog # BIRBORB138064-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse TNFRSF7 ELISA Kit
  • Detection range:
    62,5 - 4000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

Mouse TNFRSF7 ELISA Kit for the quantitative detection of mouse TNFRSF7 (CD27) in cell culture supernates and serum.

CD27 antigen, also called CD27 or TNFRSF7 is a tumour necrosis factor receptor. This gene is mapped to 12p13.31. The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is required for generation and long-term maintenance of T cell immunity. It binds to ligand CD70, and plays a key role in regulating B-cell activation and immunoglobulin synthesis. This receptor transduces signals that lead to the activation of NF-kappaB and MAPK8/JNK. Adaptor proteins TRAF2 and TRAF5 have been shown to mediate the signaling process of this receptor. CD27-binding protein (SIVA), a proapoptotic protein, can bind to this receptor and is thought to play an important role in the apoptosis induced by this receptor.