ContactUsLinkComponent
Test strips, nitrate/nitrite, Dosatest
Test strips, nitrate/nitrite, Dosatest
Catalog # 85439.601
Supplier:  VWR Chemicals
CAS Number:  
undefined
Test strips, nitrate/nitrite, Dosatest
Catalog # 85439.601
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pk:
    100
  • Description:
    Nitrate test strips 0 - 500mg/l
    Nitrite test strips 0 - 80 mg/l
  • Range:
    0 · 10 · 25 · 50 · 100 · 250 · 500
    0 · 1 · 5 · 10 · 20 · 40 · 80
Specifications
About this item
A ready to use kit that fits into your lab coat pocket.
  • Rapid and convenient
  • Easy to handle
Semi-quantitative results can be obtained between 10 and 120 seconds.
The colour charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
Packaging: Container with 100 test strips 6 x 95 mm.