Order Entry
Belgium
ContactUsLinkComponent
Mouse MER ELISA kit
Mouse MER ELISA kit
Catalog # BIRBORB138067-96
Supplier:  Biorbyt
CAS Number:  
undefined
Mouse MER ELISA kit
Catalog # BIRBORB138067-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse MER ELISA Kit
  • Detection range:
    78 - 5000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

Mouse MER ELISA Kit for the quantitative detection of mouse MER/MERTK in cell culture supernates, cell lysates, tissue homogenates and serum.

Proto-oncogene tyrosine-protein kinase MER, also called MERTK is an enzyme that in humans is encoded by the MERTK gene. This gene is a member of the MER/AXL/TYRO3 receptor kinase family and encodes a transmembrane protein with two fibronectin type-III domains, two Ig-like C2-type (immunoglobulin-like) domains, and one tyrosine kinase domain. This gene is mapped to 2q13. MERTK signaling plays a role in various processes such as macrophage clearance of apoptotic cells, platelet aggregation, cytoskeleton reorganisation and engulfment. Functions in the retinal pigment epithelium (RPE) as a regulator of rod outer segments fragments phagocytosis. This gene also plays an important role in inhibition of Toll-like receptors (TLRs)-mediated innate immune response by activating STAT1, which selectively induces production of suppressors of cytokine signaling SOCS1 and SOCS3.