Order Entry
Belgium
ContactUsLinkComponent
Human PLAT ELISA kit
Human PLAT ELISA kit
Catalog # BIRBORB138073-96
Supplier:  Biorbyt
CAS Number:  
undefined
Human PLAT ELISA kit
Catalog # BIRBORB138073-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Description:
    Human PLAT ELISA Kit
  • Detection range:
    31,2 - 2000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

Human PLAT ELISA kit for the quantitative detection of human plasminogen activator, tissue in cell culture supernates, serum, plasma (heparin, EDTA, citrate) and urine.

Plasminogen activator, tissue, also called PLAT or TPA is a serine protease. This gene encodes tissue-type plasminogen activator, a secreted serine protease which converts the proenzyme plasminogen to plasmin, a fibrinolytic enzyme. Tissue-type plasminogen activator is synthesized as a single chain which is cleaved by plasmin to a two chain disulfide linked protein. This gene was mapped to 8p11.21. This enzyme plays a role in cell migration and tissue remodeling. Increased enzymatic activity causes hyperfibrinolysis, which manifests as excessive bleeding; decreased activity leads to hypofibrinolysis which can result in thrombosis or embolism.