Order Entry
Belgium
ContactUsLinkComponent
Mouse Lipocalin-2 ELISA kit
Mouse Lipocalin-2 ELISA kit
Catalog # BIRBORB54813-96
Supplier:  Biorbyt
CAS Number:  
Mouse Lipocalin-2 ELISA kit
Catalog # BIRBORB54813-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse Lipocalin-2 ELISA Kit
  • Detection range:
    0,625 - 40 ng/ml

Specifications

About this item

Mouse Lipocalin-2 ELISA kit for the quantification of mouse lipocalin-2 in serum, plasma, urine and cell culture supernate samples.

Lipocalin-2 (LCN2), also known as neutrophil gelatinase-associated lipocalin (NGAL), 24p3, or neutrophil lipocalin (NL), is a 25-kDa secretory glycoprotein. LCN2 has been implicated in a variety of cellular processes including the innate immune response, differentiation, tumorigenesis, and cell survival.