Order Entry
Belgium
ContactUsLinkComponent
Human lipocalin-2 ELISA kit
Human lipocalin-2 ELISA kit
Catalog # BIRBORB54812-96
Supplier:  Biorbyt
CAS Number:  
Human lipocalin-2 ELISA kit
Catalog # BIRBORB54812-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Description:
    Human Lipocalin-2 ELISA Kit
  • Detection range:
    0,39 - 25 ng/ml

Specifications

About this item

Human lipocalin-2 ELISA kit for the quantification of human lipocalin-2 in serum, plasma, urine and cell culture supernate samples.

Lipocalin-2 (LCN2), also known as neutrophil gelatinase-associated lipocalin (NGAL), 24p3, or neutrophil lipocalin (NL), is a 25-kDa secretory glycoprotein. LCN2 has been implicated in a variety of cellular processes including the innate immune response, differentiation, tumorigenesis, and cell survival.

Delivery: Kit includes micro-titre strips (96 wells), wash buffer (10×40 ml), assay buffer (5×20 ml), detection antibody solution (100×0,12 ml), lyophilised human lipocalin-2 standard, substrate solution (12 ml), stop solution (12 ml).