Order Entry
Belgium
ContactUsLinkComponent
Mouse FGF-21 ELISA kit
Mouse FGF-21 ELISA kit
Catalog # BIRBORB54811-96
Supplier:  Biorbyt
CAS Number:  
Mouse FGF-21 ELISA kit
Catalog # BIRBORB54811-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse FGF-21 ELISA Kit
  • Detection range:
    31,2 - 2000 pg/ml

Specifications

About this item

Mouse FGF-21 ELISA kit is designed for the quantification of mouse fibroblast growth factor 21 (FGF-21) in serum, plasma and cell culture supernate samples.

Fibroblast growth factor 21 (FGF-21) is a novel protein that has been implicated in the regulation of lipid and glucose metabolism under fasting and ketotic conditions. In murine models, FGF-21 is predominantly expressed in liver, but it also expressed in adipose tissue and pancreatic cells. It also protects animals from diet-induced obesity when overexpressed in transgenic mice and lowers blood glucose and triglyceride levels when administered to diabetic rodents5.