Human IGFBP7 ELISA kit
Catalog # BIRBORB138093-96
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:96
- Primary antibody reactivity:Human
- Description:Human IGFBP7 ELISA Kit
- Detection range:62,5 - 40000 pg/ml
- Sensitivity:<20 pg/ml
Specifications
About this item
Human IGFBP7 ELISA kit for the quantitative detection of human insulin-like growth factor-binding protein 7 (IGFBP7) in cell culture supernates, serum, plasma (heparin, EDTA) and urine.
Insulin-like growth factor-binding protein 7 is a protein that in humans is encoded by the IGFBP7 gene. The gene was mapped to chromosome 4q12. The major function of the protein is the regulation of availability of insulin-like growth factors (IGFs) in tissue as well as in modulating IGF binding to its receptors. IGFBP7 binds to IGF with high affinity. It also stimulates cell adhesion. The protein is implicated in some cancers.
Delivery: Kit contains 96-well plate precoated with anti- human IGFBP7 antibody, lyophilised recombinant human IGFBP7 standard (2×40 ng tubes), biotinylated anti- human IGFBP7 antibody (130 µl), avidin-biotin-peroxidase complex (130 µl), sample diluent buffer (30 ml), antibody diluent buffer (12 ml), Avidin-Biotin-Peroxidase Complex diluent buffer (12 ml), TMB color developing agent (10 ml), TMB stop solution (10 ml).