Human CD200 ELISA kit
Catalog # BIRBORB138094-96
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:96
- Primary antibody reactivity:Human
- Description:Human CD200 ELISA kit, 96-well
- Detection range:31,2 - 2000 pg/ml
- Sensitivity:<10 pg/ml
Specifications
About this item
Human CD200 ELISA kit is used for quantitative detection of human CD200 in cell culture supernates, serum and plasma (heparin, EDTA).
The protein encoded by this gene is a type-1 membrane glycoprotein, which contains two immunoglobulin domains, and thus belongs to the immunoglobulin superfamily.
CD200-CD200R signaling plays a central role in macrophage fusion and osteoclast formation.