Order Entry
Belgium
ContactUsLinkComponent
Protein A detection ELISA kit
Protein A detection ELISA kit
Catalog # BIRBORB54802-1
Supplier:  Biorbyt
CAS Number:  
Protein A detection ELISA kit
Catalog # BIRBORB54802-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    HRP
  • Format:
    Pre-coated
  • Target protein:
    Staphylococcal Protein A (SpA)
  • Description:
    Protein A Detection Kit
  • Tests per kit:
    8×12 microtitre strips

Specifications

About this item

Protein A detection ELISA kit is an assay detection and miscellaneous reagent.