- Pk:96
- Primary antibody reactivity:Human
- Description:Human ALCAM ELISA kit, 96-well
- Detection range:312 - 20000 pg/ml
- Sensitivity:<10 pg/ml
Human ALCAM ELISA Kit is used for the quantitative detection of human ALCAM in cell culture supernates, serum and plasma (heparin, EDTA).
CD166 antigen is a 100 to 105 kD type I transmembrane glycoprotein that is a member of the immunoglobulin superfamily of proteins. In humans it is encoded by the ALCAM gene. The gene was mapped to human chromosome 3q13.1-q13.2 by fluorescence in situ hybridisation of cDNA probes to metaphase chromosomes. It is also called CD166, MEMD, SC-1/DM-GRASP/BEN in the chicken, and KG-CAM in the rat.
It is expressed on activated T cells, activated monocytes, epithelial cells, fibroblasts, neurons, melanoma cells, and also in sweat and sebaceous glands. CD166 protein expression is reported to be upregulated in a cell line deriving from a metastasising melanoma.
CD166 plays an important role in mediating adhesion interactions between thymic epithelial cells and CD6+ cells during intrathymic T cell development. Recently, CD166 has also been used as a potential cancer stem cell marker.