Order Entry
Belgium
ContactUsLinkComponent
Mouse MMP-12 ELISA kit
Mouse MMP-12 ELISA kit
Catalog # BIRBORB138097-96
Supplier:  Biorbyt
CAS Number:  
undefined
Mouse MMP-12 ELISA kit
Catalog # BIRBORB138097-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    MMP-12
  • Description:
    Mouse MMP-12 ELISA Kit
  • Detection range:
    62,5 - 4000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

Mouse MMP-12 ELISA kit for the quantitative detection of mouse Matrix metalloproteinase-12 in cell culture supernates, serum and plasma (heparin).

Matrix metalloproteinase-12 (MMP12), also known as MME or ME, is an enzyme that in humans is encoded by the MMP12 gene. The gene is part of a cluster of MMP genes which localise to chromosome 11q22.2. It is thought that the protein encoded by this gene is cleaved at both ends to yield the active enzyme, but this processing has not been fully described. The enzyme degrades soluble and insoluble elastin. It may play a role in aneurysm formation and studies in mice suggest a role in the development of emphysema. This gene may involved in tissue injury and remodeling.