Order Entry
Belgium
ContactUsLinkComponent
 
Human granzyme B ELISA kit
  BIRBORB138107-96
 :  Biorbyt
 :  
undefined
Human granzyme B ELISA kit
  BIRBORB138107-96
 :  Biorbyt
 :  

 

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Target protein:
    Granzyme B
  • Description:
    Human Granzyme B ELISA Kit
  • Detection range:
    15,6 - 1000 pg/ml
  • Sensitivity:
    <10 pg/ml

 

 

Human granzyme B ELISA kit for the quantitative detection of human Granzyme B in cell culture supernates, serum and plasma (heparin, EDTA).

Granzyme B is a serine protease that in humans is encoded by the GZMB gene. Granzyme B is expressed by cytotoxic T lymphocytes (CTL) and natural killer (NK) cells. CTL and NK cells share the remarkable ability to recognise specific infected target cells. They are thought to protect their host by inducing apoptosis of cells that bear on their surface nonself antigens, usually peptides or proteins resulting from infection by intracellular pathogens. The protein encoded by this gene is crucial for the rapid induction of target cell apoptosis by CTL in cell-mediated immune response.