Rat laminin ELISA kit
Catalog # BIRBORB50065-96
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:96
- Primary antibody reactivity:Rat
- Description:Rat Laminin ELISA Kit
- Detection range:156 - 10000 pg/ml
- Sensitivity:<10 pg/ml
Specifications
About this item
Rat Laminin ELISA kit for the quantitative detection of rat laminin in sera, plasma, body fluids, tissue lysates or cell culture supernates.
- No detectable cross-reactivity with other relevant proteins
Laminin is a complex extracellular glycoprotein with an approximate molecular weight of 900 kD. It plays an important role in many aspects of the cell biology. It has been found to modulate cell differentiation, cell shaping and also cell movement because it appears to be an important cell substrate-adhesion protein.
Delivery: Kit includes lyophilised recombinant rat laminin standard (2×10 ng tubes), one 96-well plate precoated with anti- rat laminin antibody, sample diluent buffer (30 ml), biotinylated anti- rat laminin antibody (130 µl), antibody diluent buffer (12 ml), avidin-biotin-peroxidase complex (ABC) (130 µl), ABC diluent buffer (12 ml), TMB color developing agent (10 ml), TMB stop solution (10 ml).