Order Entry
Belgium
ContactUsLinkComponent
Rat CD86 ELISA kit
Rat CD86 ELISA kit
Catalog # BIRBORB106612-96
Supplier:  Biorbyt
CAS Number:  
undefined
Rat CD86 ELISA kit
Catalog # BIRBORB106612-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Rat
  • Description:
    Rat CD86 ELISA kit, 96-well
  • Detection range:
    62,5 - 4000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

CD86 also known as B7-2 is a protein expressed on antigen-presenting cells that provides costimulatory signals necessary for T cell activation and survival.

The CD86 gene encodes a type I membrane protein that is a member of the immunoglobulin superfamily. Using fluorescence in situ hybridisation mapping, the CD86, like CD80, was mapped to human 3q21 and mouse chromosome 16, band B5.

The antigen presentation coactivator B72 is important in other immune-mediated thyroid diseases, lymphocytic infiltration and the immune response against thyroid carcinoma.