Specifications
- Pk:96
- Primary antibody reactivity:Rat
- Target protein:sE-selectin
- Description:Rat sE-Selectin ELISA Kit
- Detection range:156 - 10000 pg/ml
- Sensitivity:<10 pg/ml
Specifications
About this item
Rat sE-Selectin ELISA kit for the quantitative measurement of rat sE-selectin.
E-selectin, also called endothelial-leukocyte adhesion molecule-1 (ELAM-1), is a cell surface glycoprotein expressed by cytokine-activated endothelium that mediates the adhesion of blood neutrophils. An increased expression of E-selectin has been observed in the arterial endothelium interacting with lymphocytes and macrophages in human atherosclerotic lesions. E-selectin plays a critical role in mediating tissue-specific homing of T cells into skin, and of primitive hematopoietic progenitor cells (HPCs) into bone marrow (BM). A structurally and functionally related group of genes, lymph node homing receptor (LHR), granule membrane protein 140 (GMP-140), and endothelial leukocyte adhesion molecule 1 (ELAM-1) are shown to constitute a gene cluster on mouse and human chromosome 1.4.