Mouse IL-27 p28 ELISA kit
Catalog # BIRBORB106598-96
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:96
- Primary antibody reactivity:Mouse
- Target protein:IL-27 p28
- Description:Mouse IL-27 p28 ELISA Kit
- Detection range:15,6 - 1000 pg/ml
- Sensitivity:<10 pg/ml
Specifications
About this item
Mouse IL-27 p28 ELISA Kit for the quantitative determination of mouse Interleukin-27 p28.
Interleukin-27 (IL-27) is a heterodimeric cytokine belonging to the IL-12 family that is composed of two subunits; Epstein-Barr virus (EBV)-induced gene 3 (EBI3) (also known as IL-27B) and IL27-p28 (known as IL-30). IL-27 is produced by antigen-presenting cells. By genomic sequence analysis, the IL27 p28 gene was mapped to chromosome 16p11. L-27 plays an important function in regulating the activity of B- and T-lymphocytes. The effects of IL-27 are eliciting by its interaction with a specific cell-surface receptor complex composed of two proteins known as IL27R and gp130.