Order Entry
Belgium
ContactUsLinkComponent
Concanavalin A (from Jack beans), FITC (Fluorescein Isothiocyanate), MP Biomedicals
Concanavalin A (from Jack beans), FITC (Fluorescein Isothiocyanate), MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Concanavalin A (from Jack beans), FITC (Fluorescein Isothiocyanate), MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Source:
    Jack beans
  • Storage conditions:
    0 °C
  • Protein/peptide name:
    Concanavalin A
  • Concentration:
    25 mg Con A FITC/25 ml of buffer
  • Formulation:
    Frozen solution

Specifications

Frequently Bought Together