Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH tester, Eutech™ Expert pH

pH tester, Eutech™ Expert pH

Waterproof pH tester, Eutech™ Expert pH
Catalog # 663-0308
Supplier:   EUTECH
pH tester, Eutech™ Expert pH
Catalog # 663-0308
Supplier:  EUTECH
pH tester, Eutech™ Expert pH
pH tester, Eutech™ Expert pH

Waterproof pH tester, Eutech™ Expert pH

Price
114,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

This waterproof pocket pH tester is ideal for quick pH measurements in hydroponic gardening, aquaculture, agriculture, pools, education, cooling towers, environmental monitoring, water treatment also food and beverages.

  • Easy to read 4-digit dual-line display with battery life, ready and calibration indicators
  • Simultaneous display of pH and temperature
  • Automatic temperature compensation; automatic buffer recognition and calibration
  • Multi-language operational instructions
  • Upgraded key pad with intuitive interface
  • Symmetrical, leak resistant cap with a fill-line indicator and a wide, stable base

Delivery information: Supplied with four batteries and non replaceable, single junction sensor.

Documents

Specifications
  • Description:
    Waterproof pH tester, Eutech™ Expert pH
  • Compliance:
    IP 67
  • pH range:
    0,0 - 14,0
  • Power supply:
    Four batteries (A76 or LR44)
  • Temperature compensation:
    Automatic
  • pH calibration:
    Up to 3 points
  • Temperature resolution:
    0,1 °C
  • Temperature accuracy:
    ±0,5 °C + 1 least significant digit
  • Cat. No.:
    663-0308
  • pH accuracy:
    ±0,1
  • pH resolution:
    0,1
  • Model:
    Eutech™ Expert pH
  • W×D×H:
    170×45×30 mm
  • Temperature range:
    0...+50,0 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.