Mouse recombinant mRELM g (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:Store lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C.The lyophilized protein remains stable until the expiry date when stored at −20 °C.
- Protein synonyms:Resistin-like gamma
- Protein/peptide name:mRELM g
- Purity:Greater than 95% as determined by SDS-PAGE.
- Solubility:It is recommended to add 0.1M Acetate buffer pH4 to prepare working stock solution of approximately 0.5 mg/ml and let the lyophilized pellet dissolve completely. For conversion into higher pH value, we recommend intensive dilution by relevant buffer to co
- Formulation:Filtered(0.4 um) and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH-4.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together