Order Entry
Belgium
ContactUsLinkComponent
Human Leptin (from E. coli)
Human Leptin (from E. coli)
Supplier:  Biorbyt
Certificates
Human Leptin (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized Leptin although stable at room temperature, should be stored desiccated below 0 °C. Reconstituted Leptin is best stored refrigerated at 4 °C. Please avoid freeze/thaw cycles.
  • Protein synonyms:
    LEP
  • Protein/peptide name:
    Leptin
  • Purity:
    Greater than 90.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    The lyophilized Leptin is very soluble in water and most aqueous buffers below and above the isoelectric point.
  • Formulation:
    The protein was lyophilized from concentrated (1mg/ml) solution with 1X PBS, 0.1% SDS and 1mM DTT.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together