Order Entry
Belgium
Orders LinkContactUsLinkComponent
Human soluble CD25 alpha (from <i>E. coli)</i>

Human soluble CD25 alpha (from E. coli)

Marketsource
Supplier:   Biorbyt
Human soluble CD25 alpha (from <i>E. coli)</i>
Supplier:  Biorbyt
Marketsource
Certificates

2 Options Available Below

Product Details & Documents

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • BIRBORB80417-5
  • BIRBORB80417-25
  • Protein/peptide name:
  • Soluble CD25 alpha
  • Soluble CD25 alpha
  • Protein synonyms:
  • CD25
  • CD25
  • Species:
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • Purity:
  • Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Formulation:
  • IL-2R His-Tag protein is supplied in 1x PBS and 50% glycerol pH 7.4.
  • IL-2R His-Tag protein is supplied in 1x PBS and 50% glycerol pH 7.4.
  • Storage conditions:
  • Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. Please avoid freeze thaw cycles.
  • Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. Please avoid freeze thaw cycles.
  • Tested applications:
  • SDS-PAGE
  • SDS-PAGE

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 5 µG
    Log in for availability
    Log in for availability
    1 * 5 µG
    1 * 5 µG

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    BIRBORB80417-5
    Protein/peptide name
    Soluble CD25 alpha
    Protein synonyms
    CD25
    Species
    Human
    Source
    E. coli
    Purity
    Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
    Formulation
    IL-2R His-Tag protein is supplied in 1x PBS and 50% glycerol pH 7.4.
    Storage conditions
    Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. Please avoid freeze thaw cycles.
    Tested applications
    SDS-PAGE
    Marketsource
  • 25 µG
    Log in for availability
    Log in for availability
    1 * 25 µG
    1 * 25 µG

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    BIRBORB80417-25
    Protein/peptide name
    Soluble CD25 alpha
    Protein synonyms
    CD25
    Species
    Human
    Source
    E. coli
    Purity
    Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
    Formulation
    IL-2R His-Tag protein is supplied in 1x PBS and 50% glycerol pH 7.4.
    Storage conditions
    Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. Please avoid freeze thaw cycles.
    Tested applications
    SDS-PAGE
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.