Order Entry
Belgium
ContactUsLinkComponent
Human HMGB1 insect cells (from Hi-5 Insect cells)
Human HMGB1 insect cells (from Hi-5 Insect cells)
Supplier:  Biorbyt
Certificates
Human HMGB1 insect cells (from Hi-5 Insect cells)
Supplier:  Biorbyt

Specifications

  • Source:
    Hi-5 Insect cells
  • Species:
    Human
  • Storage conditions:
    HMG1 although stable 4 °C for weeks, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Protein synonyms:
    Amphoterin
  • Protein/peptide name:
    HMGB1 Insect Cells
  • Purity:
    Greater than 90% as determined by RP-HPLC and analysis by SDS-PAGE
  • Sequence:
    MGKGDPKKPR GKMSSYAFFV QTCREEHKKK HPDASVNFSE FSKKCSERWK TMSAKEKGKF EDMAKADKAR YEREMKTYIP PKGETKKKFK DPNAPKRPPS AFFLFCSEYR PKIKGEHPGL SIGDVAKKLG EMWNNTAADD KQPYEKKAAK LKEKYEKDIA AYRAKGKPDA AKKGVVKAEK SKKKKEEEED EEDEEDEEEE EDEEDEDEEE DDDDELEHHH HHH.
  • Formulation:
    The HMG1 solution (1mg/ml) contains 20 mM Tris pH-8, 1mM EDTA, 0.5mM DTT and 10% glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together