Order Entry
Belgium
ContactUsLinkComponent
Human recombinant TGFBI (from E. coli)
Human recombinant TGFBI (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant TGFBI (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Transforming Growth Factor Beta Induced protein Recombinant Human although stable at 4 °C for 30 days, should be stored desiccated below −20 °C for periods greater than 30 days. Please avoid Freeze/Thaw cycles.
  • Protein synonyms:
    Transforming growth factor-beta-induced
  • Protein/peptide name:
    TGFBI
  • Purity:
    Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Formulation:
    The TGFBI recombinant Human is formulated 20 mM Tris pH-8.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together