Order Entry
Belgium
ContactUsLinkComponent
Human recombinant P504s (from E. coli)
Human recombinant P504s (from E. coli)
Catalog # BIRBORB81249-5
Supplier:  Biorbyt
CAS Number:  
Human recombinant P504s (from E. coli)
Catalog # BIRBORB81249-5
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    5 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.
  • Protein synonyms:
    Alpha-methylacyl-CoA racemase
  • Protein/peptide name:
    P504s
  • Formulation:
    P504s at 0.1mg/ml in 50mM Tris-Acetate, pH7.5, 1mM EDTA, 20% Glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

About this item