Order Entry
Belgium
ContactUsLinkComponent
Sheep leptin triple antagonist, pegylated (from E. coli)
Sheep leptin triple antagonist, pegylated (from E. coli)
Supplier:  Biorbyt
Certificates
Sheep leptin triple antagonist, pegylated (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Sheep
  • Storage conditions:
    Lyophilized Leptin Antagonist Triple Mutant Ovine Recombinant although stable at room temperature for several weeks, should be stored desiccated below −18 °C. Upon reconstitution at 0. 1 Lep-tA mutant mg/ml and up to 2mM and filter sterilization Leptin mutant can be stored at 4 °C or even room temperature for several weeks making it suitable for long term infusion studies using osmotic pumps. At lower concentration addition of carrier protein (0. 1% HSA or BSA) is suggested. Please prevent freeze/thaw cycles.
  • Biological activity:
    Pegylated triple antagonist is capable of inhibiting leptin-induced proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. Its in vitro activity is 6-8 fold lower than the non-pegylated antagonist but in vivo it has profound weight gain effect (as compared to the non-pegylated antagonist like in mouse leptin antagonists), resulting mainly from increased food intake.
  • Protein synonyms:
    LEP
  • Protein/peptide name:
    Leptin Triple Antagonist, pegylated
  • Purity:
    Greater than 95.0% as determined by: (a) Gel filtration analysis. (b) Analysis by SDS-PAGE.
  • Solubility:
    It is recommended to reconstitute the lyophilized Leptin-Antagonist Triple Mutant Ovine Recombinant in sterile water or sterile 0.4% NaHCO3 adjusted to pH 8, not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Formulation:
    The protein was lyophilized from concentrated (0.65mg/ml) solution with 0.003mM NaHCO3.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together