Order Entry
Belgium
ContactUsLinkComponent
Mouse Leptin, pegylated (from E. coli)
Mouse Leptin, pegylated (from E. coli)
Supplier:  Biorbyt
Certificates
Mouse Leptin, pegylated (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Lyophilized Leptin although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution Leptin should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Biological activity:
    Pegylated mouse Leptin is capable of stimulatng proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. Pegylated mouse Leptin in vitro activity is only slightly lower than the non-pegylated antagonist but in vivo it has profound weight reducing effect (as compared to the non-pegylated leptin), resulting mainly from reduced food intake.
  • Protein synonyms:
    LEP
  • Protein/peptide name:
    Leptin, pegylated
  • Purity:
    Greater than 99.0% as determined by(a) Analysis by Gel-Filtration.(b) Analysis by SDS-PAGE.
  • Solubility:
    It is recommended to reconstitute the lyophilized Leptin in sterile 0.4% NaHCO3 adjusted to pH-8.5 not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Formulation:
    The mouse Leptin was lyophilized from concentrated (0.65mg/ml) solution containing 0.003mM NaHCO3.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together