Order Entry
Belgium
ContactUsLinkComponent
Mouse Leptin triple antagonist, pegylated (from E. coli)
Mouse Leptin triple antagonist, pegylated (from E. coli)
Supplier:  Biorbyt
Certificates
Mouse Leptin triple antagonist, pegylated (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Lyophilized Leptin Antagonist Triple Mutant Mouse Recombinant although stable at room temperature for several weeks, should be stored desiccated below −18 °C. Upon reconstitution at 0. 1 Leptin mutant mg/ml and up to 2mM and filter sterilization LEP mutant can be stored at 4 °C or even room temperature for several weeks making it suitable for long term infusion studies using osmotic pumps. At lower concentration addition of carrier protein (0. 1% HSA or BSA) is suggested. Please prevent freeze/thaw cycles.
  • Biological activity:
    Leptin Antagonist Triple Mutant Mouse Recombinant half-life in circulation after SC injection was over 20 hours.Leptin Antagonist Triple Mutant Mouse Recombinant is capable of inhibiting leptin-induced proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. Leptin Antagonist Triple Mutant Mouse Recombinant in vitro activity is 5-6 fold lower than the non-pegylated antagonist, though in vivo it has profound weight gain effect (as compared to the non-pegylated antagonist), resulting mainly from increased food intake.
  • Protein synonyms:
    LEP
  • Protein/peptide name:
    Leptin Triple Antagonist, Pegylated
  • Purity:
    Greater than 99.0% as determined by(a) Gel filtration analysis.(b) Analysis by SDS-PAGE.
  • Solubility:
    It is recommended to reconstitute the lyophilized Leptin Antagonist Triple Mutant Mouse Recombinant in sterile water or sterile 0.4% NaHCO3adjusted to pH 8-9, not less than 100ug/ml, which can then be further diluted with other aqueous solutions.
  • Formulation:
    The The Mouse Leptin triple anatagonist was lyophilized from concentrated (0.65mg/ml) solution with 0.003mM NaHCO3.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together