Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Midkine (from E. coli)
Human recombinant Midkine (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant Midkine (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C.The lyophilized protein remains stable until the expiry date when stored at −20 °C.
  • Protein synonyms:
    NEGF-2
  • Protein/peptide name:
    Midkine
  • Purity:
    Greater than 95% as determined by SDS-PAGE. Three-step procedure using affinity Ni-NTA chromatography and size exclusion chromatography before and after refolding.
  • Solubility:
    Add 0.2 uml of PBS pH 7.2 and let the lyophilized pellet dissolve completely.
  • Formulation:
    Lyophilized from 0.5mg/ml in 0.05M phosphate buffer and 0.1M NaCl, pH 7.2.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together