Order Entry
Belgium
ContactUsLinkComponent
Human IL4 (from E. coli)
Human IL4 (from E. coli)
Catalog # BIRBORB80061-5
Supplier:  Biorbyt
CAS Number:  
Human IL4 (from E. coli)
Catalog # BIRBORB80061-5
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    5 µG
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    The ED50 as determined by the dose-dependant stimulation of TF-1 cells is 0.2 ng/ml, corresponding to Specific Activity of 5x1,000,000 IU/mg.
  • Protein synonyms:
    B cell growth factor 1
  • Protein/peptide name:
    IL4
  • Purity:
    > 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Interleukin-4 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    The sequence of the first five N-terminal amino acids was determined and was found to be Met-His-Lys-Cys-Asp.
  • Formulation:
    Lyophilized from concentrated (1mg/ml) solution in water containing no additives.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

About this item