Order Entry
Belgium
ContactUsLinkComponent
Zea mays CK alpha (from Escherichia coli)
Zea mays CK alpha (from Escherichia coli)
Supplier:  Biorbyt
Certificates
Zea mays CK alpha (from Escherichia coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Zea mays
  • Storage conditions:
    Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
  • Protein synonyms:
    EC 27111
  • Protein/peptide name:
    CK alpha
  • Purity:
    Greater than 99% as determined by SDS-PAGE.
  • Formulation:
    CK2a is supplied in 50% glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together