Order Entry
Belgium
ContactUsLinkComponent
Mycobacterium Tuberculosis major secretory Antigen 85B (from E. coli)
Mycobacterium Tuberculosis major secretory Antigen 85B (from E. coli)
Supplier:  Biorbyt
Certificates
Mycobacterium Tuberculosis major secretory Antigen 85B (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Storage conditions:
    Ag85B although stable room temperature for weeks, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA).Please prevent freeze-thaw cycles.
  • Protein synonyms:
    Antigen 85-B
  • Protein/peptide name:
    Mycobacterium Tuberculosis major secretory Antigen 85B
  • Purity:
    Greater than 90.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Antigen-85B in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Formulation:
    The protein was lyophilized with no additives.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together