Order Entry
Belgium
ContactUsLinkComponent
 
Mouse recombinant KGF (from E. coli)
  BIRBORB168697-2
 :  Biorbyt
 :  
Mouse recombinant KGF (from E. coli)
  BIRBORB168697-2
 :  Biorbyt
 :  

 

  • Pk:
    2 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Store at 4 °C for short term and at −20 °C for long term storage
  • Protein synonyms:
    HBGF-7
  • Protein/peptide name:
    KGF
  • Purity:
    > 95.0% purified
  • Sequence:
    MCNDMSPEQT ATSVNCSSPE RHTRSYDYME GGDIRVRRLF CRTQWYLRID KRGKVKGTQE MKNSYNIMEI RTVAVGIVAI KGVESEYYLA MNKEGKLYAK KECNEDCNFK ELILENHYNT YASAKWTHSG GEMFVALNQK GIPVKGKKTK KEQKTAHFLP MAIT.
  • Formulation:
    20mM Phosphate buffer pH-8 and 0.1M NaCl.
  • Tested applications:
    SDS-PAGE