Order Entry
Belgium
ContactUsLinkComponent
Mouse Bax (from E. coli)
Mouse Bax (from E. coli)
Supplier:  Biorbyt
Certificates
Mouse Bax (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Mouse Bax although stable at 12 °C for week, should be stored desiccated below −18 °C. Please prevent freeze/thaw cycles.
  • Protein synonyms:
    Apoptosis regulator BAX membrane isoform alpha
  • Protein/peptide name:
    Bax
  • Purity:
    Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Formulation:
    The protein contains 10mM Tris-HCL pH-8, 1mM EDTA and 250mM NaCl.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together