Mouse mBID GST (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Mouse
- Storage conditions:GST BID although stable at 15 °C for week, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Protein/peptide name:mBID GST
- Purity:Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Formulation:The Mouse GST tag BID protein solution contains 10mM Tris-HCL pH-8, 1mM EDTA and 250mM NaCl.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together