Order Entry
Belgium
ContactUsLinkComponent
Human recombinant VSNL1 His (from E. coli)
Human recombinant VSNL1 His (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant VSNL1 His (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/ thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time.
  • Protein synonyms:
    VISL1
  • Protein/peptide name:
    VSNL1 His
  • Purity:
    Greater than 95% as determined by SDS-PAGE.
  • Solubility:
    Add sterile deionized water to working concentration of 0.5mg/ml and let the lyophilized pellet dissolve completely.
  • Sequence:
    MKHHHHHHAS MGKQNSKLAP EVMEDLVKST EFNEHELKQW YKGFLKDCPS GRLNLEEFQQ LYVKFFPYGD ASKFAQHAFR TFDKNGDGTI DFREFICALS ITSRGSFEQK LNWAFNMYDL DGDGKITRVE MLEIIEAIYK MVGTVIMMKM NEDGLTPEQR VDKIFSKMDK NKDDQITLDE FKEAAKSDPS IVLLLQCDIQK.
  • Formulation:
    The sterile filtered concentrated (0.5mg/ml) protein solution was lyophilized with 20 mM Tris 20 mM NaCl pH-7.5.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together