Human tissue factor (from E. coli)
BIRBORB81422-2
: Biorbyt
:
- Pk:2 µG
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. Please avoid freeze thaw cycles.
- Protein synonyms:CD 142
- Protein/peptide name:Tissue Factor
- Purity:Greater than 95.0% as determined by SDS-PAGE.
- Formulation:Tissue factor protein is supplied in 22mM Tris.HCl pH8.0, 1mM EDTA and 50% glycerol.
- Tested applications:SDS-PAGE