Order Entry
Belgium
ContactUsLinkComponent
Human VEGF receptor 1 (from E. coli)
Human VEGF receptor 1 (from E. coli)
Supplier:  Biorbyt
Certificates
Human VEGF receptor 1 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. Please avoid freeze thaw cycles.
  • Protein synonyms:
    Vascular endothelial growth factor A
  • Protein/peptide name:
    VEGF receptor 1
  • Purity:
    Greater than 95.0% as determined by SDS-PAGE.
  • Formulation:
    FLT1 His-Tag protein is supplied in 1x PBS and 50% glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together