Order Entry
Belgium
ContactUsLinkComponent
Human recombinant PKC-g (from insect cells (Sf9))
Human recombinant PKC-g (from insect cells (Sf9))
Fournisseur:  Biorbyt
Certificats
Human recombinant PKC-g (from insect cells (Sf9))
Fournisseur:  Biorbyt

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells (Sf9)
  • Species:
    Human
  • Storage conditions:
    Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
  • Protein synonyms:
    Protein kinase gamma type
  • Protein/peptide name:
    PKC-g
  • Purity:
    Greater than 90% as determined by SDS-PAGE.
  • Formulation:
    PKC-g is supplied at of 0.1mg/ml in 10mM Tris, pH 7.4, 0.1M NaCl, 20% glycerol, 1mM DTT, 0.1mM EDTA, 0.2 um PMSF and 0.03% Brij-35.
  • Tested applications:
    SDS-PAGE

Spécifications

Frequently Bought Together