Human recombinant Resistin Mutant (from E. coli)
: Biorbyt
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C.The lyophilized protein remains stable until the expiry date when stored at −20 °C.
- Protein synonyms:Resistin
- Protein/peptide name:Resistin Mutant
- Purity:Greater than 95% as determined by SDS-PAGE.
- Solubility:Add 0.2 uml of deionized water and let the lyophilized pellet dissolve completely.
- Formulation:Recombinant Human Resistin was lyophilized from concentrated (1mg/ml) solution containing 0.1% TFA.
- Tested applications:SDS-PAGE
Frequently Bought Together