Order Entry
Belgium
ContactUsLinkComponent
Human recombinant CLU (from Plasma)
Human recombinant CLU (from Plasma)
Supplier:  Biorbyt
Certificates
Human recombinant CLU (from Plasma)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Plasma
  • Species:
    Human
  • Storage conditions:
    Store lyophilized Clusterin at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C. The lyophilized protein remains stable until the expiry date when stored at −20 °C.
  • Protein synonyms:
    Clusterin
  • Protein/peptide name:
    CLU
  • Purity:
    Greater than 95% as determined by SDS PAGE.
  • Solubility:
    Add deionized H2O to working volume of 0.5 mg/ml and let the lyophilized pellet dissolve completely.
  • Formulation:
    Human native Clusterin was lyophilized from PBS, pH 7.5.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together