Human recombinant IL7 His (from E. coli)
: Biorbyt
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. Please avoid freeze/thaw cycles.
- Protein synonyms:LP-1|Lymphopoietin
- Protein/peptide name:IL7 His
- Purity:Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Formulation:Interleukin -7 His is supplied in 1x PBS and 50% glycerol.
- Tested applications:SDS-PAGE
Frequently Bought Together