Human adiponectin glycosilated HEK (from HEK293 cells)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:HEK293 cells
- Species:Human
- Storage conditions:For long term, store lyophilized AdipoQ at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C. The lyophilized protein remains stable for 24 months when stored at −20 °C.
- Biological activity:In vitro gluconeogenesis assay in primary hepatocytes was performed, showing the Adiponectin human derived from mammalian cells can inhibit glucose production. The ED50 was ~6 ug/ml.
- Protein synonyms:ACDC
- Protein/peptide name:Adiponectin glycosilated HEK
- Purity:Greater than 98% as determined by HPLC and SDS PAGE.
- Solubility:Add 0.2 uml of deionized water and let the lyophilized pellet dissolve completely.
- Sequence:ETTTQGPGVL LPLPKGACTG WMAGIPGHPG HNGAPGRDGR DGTPGEKGEK GDPGLIGPKG DIGETGVPGA EGPRGFPGIQ GRKGEPGEGA YVYRSAFSVG LETYVTIPNM PIRFTKIFYN QQNHYDGSTG KFHCNIPGLY YFAYHIVYMK DVKVSLFKKD KAMLFTYDQY QENNVDQASG SVLLHLEVGD QVWLQVYGEG ERNGLYADND NDSTFTGFLL YHDTNDYKDDDDK.
- Formulation:Sterile filtered and lyophilized from 0.5 mg/ml in PBS buffer.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together