Order Entry
Belgium
ContactUsLinkComponent
Human recombinant GM CSF Pichia (from P. pastoris)
Human recombinant GM CSF Pichia (from P. pastoris)
Supplier:  Biorbyt
Certificates
Human recombinant GM CSF Pichia (from P. pastoris)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    P. pastoris
  • Species:
    Human
  • Storage conditions:
    Lyophilized Granulocyte Macrophage Colony Stimulating Factor although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution GMCSF should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Biological activity:
    The ED50 as determined by the dose-dependant stimulation of the proliferation of human TF-1 cells (human erythroleukemic indicator cell line) is 0.183 ng/ml, corresponding to Specific Activity of 5,000,000 IU/mg.
  • Protein synonyms:
    CSF-2
  • Protein/peptide name:
    GM CSF Pichia
  • Purity:
    Greater than 97.0% as determined by1. Analysis by RP-HPLC.2. Analysis by SDS-PAGE.
  • Solubility:
    It is recommended to reconstitute the lyophilized Granulocyte Macrophage Colony Stimulating Factor in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    The sequence of the first five N-terminal amino acids was determined and was found to be Ala-Pro-Ala-Arg-Ser.
  • Formulation:
    The protein was lyophilized from concentrated (1mg/ml) solution containing 10mM phosphate buffer pH 7.0, 40 mg mannitol and 10 mg sucrose.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together