Order Entry
Belgium
ContactUsLinkComponent
Human adiponectin (globular) (from E. coli)
Human adiponectin (globular) (from E. coli)
Supplier:  Biorbyt
Certificates
Human adiponectin (globular) (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    For long term, store lyophilized Acrp30 Human at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted Acrp30 Human can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C. The lyophilized Acrp30 Human remains stable for 24 months when stored at −20 °C.
  • Gene ID:
    9370
  • Protein synonyms:
    ACDC
  • Protein/peptide name:
    Adiponectin (globular)
  • Purity:
    Purity of Acrp30 Human is greater than 95% as determined by HPLC and SDS PAGE.
  • Solubility:
    Add 0.2 uml of deionized water and let the lyophilized pellet of Acrp30 Human dissolve completely.
  • Sequence:
    MSWWHHHHHH NWNIPTTQDT TQDLWFEGAM GGEGAYVYRS FSVGLETYV TIPNMPIRFT KIFYNQQNHYDGSTGKFHCN IPGLYYFAYH ITVYMKDVKV SLFKKDKAML FTYDQYQENN VDQASGSVLL HEVGDQVWLQVYGEGERNGL YADNDNDSTF TGFLLYHDTN (Swiss-prot entry Q15848).
  • Formulation:
    Acrp30 Human is filtered powder, lyophilized from 0.6 mg/ml in PBS buffer.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together