Order Entry
Belgium
ContactUsLinkComponent
Rat VEGF related (from insect cells (Sf9))
Rat VEGF related (from insect cells (Sf9))
Supplier:  Biorbyt
Certificates
Rat VEGF related (from insect cells (Sf9))
Supplier:  Biorbyt

Specifications

  • Source:
    Insect cells (Sf9)
  • Species:
    Rat
  • Storage conditions:
    Lyophilized Vascular Endothelial Growth Factor-C although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution VEGF-C should be stored at 4 °C between 2-7 days and for future use below −18 °C. Please prevent freeze/thaw cycles.
  • Biological activity:
    Measured by its ability to stimulate phosphorylation of the VEGFR-3/FLT-4 receptor in porcine aortic endothelial cells. The ED50 for this effect is typically 200-300 ng/ml.
  • Protein synonyms:
    Vascular endothelial growth factor A
  • Protein/peptide name:
    VEGF Related
  • Purity:
    Greater than 90.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Vascular Endothelial Growth Factor in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Formulation:
    Each mg of VEGF-C Rat contains 50mg BSA and PBS as buffer.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together